Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLF13
Entrez Gene ID
101110272
Gene Name
Kruppel like factor 13
Gene Aliases
-
Chromosome Location
Chr.18: 25552226 - 25590011 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018077.3 XP_004018126.2 1 - 2 966 106

View Full Product Details