Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS7BP
Entrez Gene ID
101105358
Gene Name
regulator of G protein signaling 7 binding protein
Gene Aliases
-
Chromosome Location
Chr.16: 15293630 - 15403926 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016942.4 XP_004016991.1 4 - 5 1299 110

View Full Product Details