Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCN1
Entrez Gene ID
101111999
Gene Name
transcobalamin 1
Gene Aliases
-
Chromosome Location
Chr.15: 89377645 - 89392162 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012096487.3 XP_011951877.2 9 - 10 1387 115

View Full Product Details