Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H12B
Entrez Gene ID
101112028
Gene Name
zinc finger CCCH-type containing 12B
Gene Aliases
-
Chromosome Location
Chr.X: 49460943 - 49640211 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015104870.2 XP_014960356.1 3 - 4 1770 97

View Full Product Details