Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM4
Entrez Gene ID
101111684
Gene Name
tripartite motif containing 4
Gene Aliases
-
Chromosome Location
Chr.24: 37155314 - 37170991 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021042.4 XP_004021091.1 5 - 6 1155 93

View Full Product Details