Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD37
Entrez Gene ID
101112948
Gene Name
ankyrin repeat domain 37
Gene Aliases
-
Chromosome Location
Chr.26: 16625549 - 16642194 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012105729.3 XP_011961119.1 4 - 5 690 105

View Full Product Details