Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL19
Entrez Gene ID
101112663
Gene Name
interleukin 19
Gene Aliases
-
Chromosome Location
Chr.12: 4238249 - 4247654 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012186967.3 XP_012042357.1 3 - 4 258 105

View Full Product Details