Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GNRH2P
Entrez Gene ID
101112753
Gene Name
gonadotropin releasing hormone 2 pseudogene
Gene Aliases
-
Chromosome Location
Chr.13: 54232316 - 54237852 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_160973.1 1 - 2 181 108

View Full Product Details