Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PROSER2
Entrez Gene ID
101116497
Gene Name
proline and serine rich 2
Gene Aliases
C13H10orf47
Chromosome Location
Chr.13: 16652953 - 16694718 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004014184.4 XP_004014233.2 2 - 3 290 97

View Full Product Details