Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RTL9
Entrez Gene ID
101113570
Gene Name
retrotransposon Gag like 9
Gene Aliases
RGAG1
Chromosome Location
Chr.X: 133234569 - 133240711 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963256.1 XP_027819057.1 1 - 2 4490 111

View Full Product Details