Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF6
Entrez Gene ID
101115804
Gene Name
ring finger protein 6
Gene Aliases
-
Chromosome Location
Chr.10: 35045446 - 35055179 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015098175.2 XP_014953661.2 3 - 4 488 96
XM_027973647.1 XP_027829448.1 3 - 4 441 96
XM_027973648.1 XP_027829449.1 3 - 4 441 96
XM_027973649.1 XP_027829450.1 3 - 4 388 96
XM_027973650.1 XP_027829451.1 3 - 4 387 96
XM_027973651.1 XP_027829452.1 3 - 4 397 96

View Full Product Details