Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRP1
Entrez Gene ID
101114035
Gene Name
NLR family pyrin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.11: 37300303 - 37332571 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012185551.3 XP_012040941.2 10 - 11 3247 79
XM_027975093.1 XP_027830894.1 9 - 10 3055 79
XM_027975095.1 XP_027830896.1 10 - 11 3099 79

View Full Product Details