Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HHEX
Entrez Gene ID
100008709
Gene Name
hematopoietically expressed homeobox
Gene Aliases
HEX
Chromosome Location
Chr.18: 40478044 - 40483975 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002718509.3 XP_002718555.1 2 - 3 614 65
GenBank mRNA AF102133.1 - 1 - 2 178 65

View Full Product Details