The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_002711632.3 | XP_002711678.1 | 1 - 2 | 405 | 151 |
XM_008257512.2 | XP_008255734.1 | 1 - 2 | 406 | 151 | |
XM_017342314.1 | XP_017197803.1 | 1 - 2 | 406 | 151 | |
GenBank mRNA | AJ291311.1 | - | 1 - 2 | 72 | 151 |