Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
F2
Entrez Gene ID
100009146
Gene Name
coagulation factor II, thrombin
Gene Aliases
THROMBIN
Chromosome Location
Chr.1: 186084649 - 186102085 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002709082.3 XP_002709128.1 10 - 11 1325 105
GenBank mRNA M81396.1 - 1 - 2 146 105

View Full Product Details