Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALOX5AP
Entrez Gene ID
100009413
Gene Name
arachidonate 5-lipoxygenase activating protein
Gene Aliases
FLAP
Chromosome Location
Chr.Unknown: 1158024 - 1187525 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002720606.3 XP_002720652.1 - 162 67
GenBank mRNA M96556.1 - - 73 67

View Full Product Details