Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC4A9
Entrez Gene ID
100008621
Gene Name
solute carrier family 4 member 9
Gene Aliases
AE4A
Chromosome Location
Chr.3: 22495679 - 22534757 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082005.1 NP_001075474.1 12 - 13 1710 64
XM_008254828.2 XP_008253050.1 12 - 13 2260 64
XM_008254829.2 XP_008253051.1 12 - 13 2309 64
XM_017340967.1 XP_017196456.1 12 - 13 2309 64
XM_017340968.1 XP_017196457.1 12 - 13 2310 64
XM_017340969.1 XP_017196458.1 12 - 13 2261 64
XM_017340970.1 XP_017196459.1 12 - 13 2241 64
XM_017340971.1 XP_017196460.1 11 - 12 2195 64
XM_017340972.1 XP_017196461.1 12 - 13 2193 64
XM_017340974.1 XP_017196463.1 11 - 12 1465 64
XM_017340975.1 XP_017196464.1 12 - 13 2312 64
GenBank mRNA AB038263.1 - 12 - 13 1710 64
AB038264.1 - 12 - 13 1662 64

View Full Product Details