Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC4A9
Entrez Gene ID
100008621
Gene Name
solute carrier family 4 member 9
Gene Aliases
AE4A
Chromosome Location
Chr.3: 22495679 - 22534757 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082005.1 NP_001075474.1 5 - 6 720 98
XM_008254828.2 XP_008253050.1 5 - 6 1318 98
XM_008254829.2 XP_008253051.1 5 - 6 1319 98
XM_017340967.1 XP_017196456.1 5 - 6 1319 98
XM_017340968.1 XP_017196457.1 5 - 6 1320 98
XM_017340969.1 XP_017196458.1 5 - 6 1319 98
XM_017340970.1 XP_017196459.1 5 - 6 1320 98
XM_017340971.1 XP_017196460.1 4 - 5 1205 98
XM_017340972.1 XP_017196461.1 5 - 6 1320 98
XM_017340973.1 XP_017196462.1 5 - 6 1322 98
XM_017340974.1 XP_017196463.1 4 - 5 475 98
XM_017340975.1 XP_017196464.1 5 - 6 1322 98
GenBank mRNA AB038263.1 - 5 - 6 720 98
AB038264.1 - 5 - 6 720 98

View Full Product Details