Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITIH3
Entrez Gene ID
100008624
Gene Name
inter-alpha-trypsin inhibitor heavy chain 3
Gene Aliases
ITI-HC3
Chromosome Location
Chr.9: 19828876 - 19841123 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082008.1 NP_001075477.1 8 - 9 907 114
XM_008260447.2 XP_008258669.1 8 - 9 936 114
GenBank mRNA AB050593.1 - 8 - 9 907 114

View Full Product Details