Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
NDEL1
Entrez Gene ID
100008657
Gene Name
nudE neurodevelopment protein 1 like 1
Gene Aliases
-
Chromosome Location
Chr.19: 10830833 - 10863389 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082020.1 NP_001075489.1 - 1111 64
GenBank mRNA AF015037.3 - - 1111 64

View Full Product Details