Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NR1I2
Entrez Gene ID
100008739
Gene Name
nuclear receptor subfamily 1 group I member 2
Gene Aliases
PXR
Chromosome Location
Chr.14: 98968609 - 99000849 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082067.2 NP_001075536.1 8 - 9 1346 77
GenBank mRNA AF182217.1 - 8 - 9 1347 77

View Full Product Details