Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VCAM1
Entrez Gene ID
100008901
Gene Name
vascular cell adhesion molecule 1
Gene Aliases
VCAM-1
Chromosome Location
Chr.13: 63440580 - 63462573 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082152.1 NP_001075621.1 8 - 9 2163 124
GenBank mRNA AY212510.1 - 8 - 9 2163 124

View Full Product Details