Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADCY10
Entrez Gene ID
100008902
Gene Name
adenylate cyclase 10
Gene Aliases
SAC
Chromosome Location
Chr.13: 25099914 - 25203191 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082153.1 NP_001075622.1 32 - 33 4969 73
XM_008263960.2 XP_008262182.1 32 - 33 5323 73
GenBank mRNA AY212921.1 - 32 - 33 4969 73

View Full Product Details