Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC36A1
Entrez Gene ID
100008971
Gene Name
solute carrier family 36 member 1
Gene Aliases
PAT1, Slc36a3
Chromosome Location
Chr.3: 32917899 - 32969579 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082183.1 NP_001075652.1 8 - 9 933 104
XM_008254838.2 XP_008253060.1 8 - 9 1012 104
XM_008254845.2 XP_008253067.1 8 - 9 1127 104
XM_008254846.2 XP_008253068.1 8 - 9 1046 104
XM_017340981.1 XP_017196470.1 8 - 9 1023 104
GenBank mRNA AY989816.1 - 8 - 9 933 104

View Full Product Details