Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KITLG
Entrez Gene ID
100009028
Gene Name
KIT ligand
Gene Aliases
Kitl
Chromosome Location
Chr.4: 67503403 - 67541344 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082223.1 NP_001075692.1 3 - 4 356 130
GenBank mRNA DQ356265.1 - 3 - 4 356 130

View Full Product Details