Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LGALS3
Entrez Gene ID
100009187
Gene Name
galectin 3
Gene Aliases
galig
Chromosome Location
Chr.17: 73821809 - 73827372 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082338.1 NP_001075807.1 4 - 5 593 86
GenBank mRNA U06470.1 - 4 - 5 593 86

View Full Product Details