Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EEF1A1
Entrez Gene ID
100009189
Gene Name
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases
RABEFLA2
Chromosome Location
Chr.12: 60379747 - 60382266 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082339.1 NP_001075808.1 6 - 7 1313 78
GenBank mRNA U09823.1 - 6 - 7 1313 78

View Full Product Details