Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLMAP
Entrez Gene ID
100009199
Gene Name
sarcolemma associated protein
Gene Aliases
SLAP
Chromosome Location
Chr.9: 24582901 - 24751719 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082348.2 NP_001075817.2 18 - 19 2041 75
XM_008260454.2 XP_008258676.1 21 - 22 3379 75
XM_008260455.2 XP_008258677.1 21 - 22 3373 75
XM_008260456.2 XP_008258678.1 21 - 22 3379 75
XM_008260457.2 XP_008258679.1 20 - 21 3328 75
XM_008260458.2 XP_008258680.1 20 - 21 3328 75
XM_008260459.2 XP_008258681.1 20 - 21 3322 75
XM_008260460.2 XP_008258682.1 20 - 21 3316 75
XM_008260461.2 XP_008258683.1 20 - 21 3310 75
XM_008260462.2 XP_008258684.1 20 - 21 3310 75
XM_008260463.2 XP_008258685.1 19 - 20 3277 75
XM_008260464.2 XP_008258686.1 19 - 20 3265 75
XM_008260465.2 XP_008258687.1 19 - 20 3265 75
XM_008260466.2 XP_008258688.1 20 - 21 3258 75
XM_008260467.2 XP_008258689.1 18 - 19 3213 75
XM_008260468.2 XP_008258690.1 18 - 19 3207 75
XM_008260470.2 XP_008258692.1 18 - 19 3156 75
XM_008260471.2 XP_008258693.1 18 - 19 3149 75
XM_008260472.2 XP_008258694.1 17 - 18 3092 75
XM_008260473.2 XP_008258695.1 17 - 18 3086 75
XM_017343638.1 XP_017199127.1 19 - 20 3271 75
XM_017343639.1 XP_017199128.1 21 - 22 3384 75
XM_017343640.1 XP_017199129.1 7 - 8 920 75
XM_017343641.1 XP_017199130.1 7 - 8 941 75
XM_017343642.1 XP_017199131.1 6 - 7 799 75
XM_017343643.1 XP_017199132.1 5 - 6 677 75
GenBank mRNA AF304449.1 - 7 - 8 882 75
U21155.1 - 6 - 7 759 75
U21156.1 - 7 - 8 831 75
U21157.1 - 17 - 18 2279 75

View Full Product Details