Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMED10
Entrez Gene ID
100009296
Gene Name
transmembrane p24 trafficking protein 10
Gene Aliases
-
Chromosome Location
Chr.20: 29940963 - 29994129 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082408.1 NP_001075877.1 3 - 4 454 63
XM_008271895.2 XP_008270117.1 3 - 4 469 63
GenBank mRNA X98303.1 - - 454 63

View Full Product Details