Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAS3
Entrez Gene ID
100009454
Gene Name
hyaluronan synthase 3
Gene Aliases
-
Chromosome Location
Chr.5: 24516758 - 24529437 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082709.1 NP_001076178.1 2 - 3 733 81
XM_008257539.2 XP_008255761.1 3 - 4 1194 81
XM_017342182.1 XP_017197671.1 3 - 4 1342 81
GenBank mRNA AB055979.1 - 2 - 3 733 81

View Full Product Details