Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCNN1A
Entrez Gene ID
100009488
Gene Name
sodium channel epithelial 1 subunit alpha
Gene Aliases
ALPHA-ENAC
Chromosome Location
Chr.8: 32419340 - 32443570 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082728.1 NP_001076197.1 10 - 11 1459 72
GenBank mRNA AJ132108.1 - 10 - 11 1459 72
AY521461.1 - 9 - 10 1442 72

View Full Product Details