Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITIH2
Entrez Gene ID
100009345
Gene Name
inter-alpha-trypsin inhibitor heavy chain 2
Gene Aliases
ITI-HC2
Chromosome Location
Chr.16: 12056544 - 12100378 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082647.2 NP_001076116.1 18 - 19 2511 134
XM_008268046.2 XP_008266268.1 17 - 18 2517 134
GenBank mRNA AB050592.1 - 18 - 19 2513 134

View Full Product Details