Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC9A3
Entrez Gene ID
100009430
Gene Name
solute carrier family 9 member A3
Gene Aliases
NHE-3
Chromosome Location
Chr.Unknown: 226309 - 237120 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082697.1 NP_001076166.1 - 1562 63
GenBank mRNA M87007.1 - - 1562 63

View Full Product Details