Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LGALS4
Entrez Gene ID
100009464
Gene Name
galectin 4
Gene Aliases
-
Chromosome Location
Chr.5: 949711 - 956803 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082713.1 NP_001076182.1 - 476 87
GenBank mRNA AF091738.1 - - 476 87

View Full Product Details