Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRO
Entrez Gene ID
100009529
Gene Name
protein tyrosine phosphatase receptor type O
Gene Aliases
GLEPP1, PTP
Chromosome Location
Chr.8: 23969041 - 24227954 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082750.1 NP_001076219.1 7 - 8 1639 100
XM_008259342.2 XP_008257564.1 7 - 8 1678 100
XM_017343246.1 XP_017198735.1 7 - 8 1591 100
GenBank mRNA U09490.1 - 7 - 8 1639 100

View Full Product Details