Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC28A2
Entrez Gene ID
100009595
Gene Name
solute carrier family 28 member 2
Gene Aliases
SPNT1, Slc28a1
Chromosome Location
Chr.17: 27287589 - 27320640 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082791.1 NP_001076260.1 6 - 7 647 64
GenBank mRNA AF161716.1 - 6 - 7 647 64

View Full Product Details