Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFIP11
Entrez Gene ID
100101578
Gene Name
tuftelin interacting protein 11
Gene Aliases
STIP
Chromosome Location
Chr.Unknown: 127706 - 142639 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099965.1 NP_001093435.1 - 805 57
XM_008274606.2 XP_008272828.1 - 910 57
XM_008274607.2 XP_008272829.1 - 1128 57
XM_017350649.1 XP_017206138.1 1 - 1 565 57
GenBank mRNA EF443269.1 - - 805 57

View Full Product Details