Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TFIP11
Entrez Gene ID
100101578
Gene Name
tuftelin interacting protein 11
Gene Aliases
STIP
Chromosome Location
Chr.Unknown: 127706 - 142639 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099965.1 NP_001093435.1 - 1856 97
XM_008274606.2 XP_008272828.1 - 1961 97
XM_008274607.2 XP_008272829.1 - 2179 97
XM_017350649.1 XP_017206138.1 1 - 1 1616 97
GenBank mRNA EF443269.1 - - 1856 97

View Full Product Details