Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYH4
Entrez Gene ID
100125991
Gene Name
myosin heavy chain 1
Gene Aliases
MHC, MYH1
Chromosome Location
Chr.19: 9056212 - 9080806 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109816.1 NP_001103286.1 37 - 38 5732 137
GenBank mRNA U32574.1 - 37 - 38 5732 137

View Full Product Details