Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL18R1
Entrez Gene ID
100303768
Gene Name
interleukin 18 receptor 1
Gene Aliases
-
Chromosome Location
Chr.2: 90029967 - 90075973 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164863.1 NP_001158335.1 4 - 5 621 96
XM_008252716.2 XP_008250938.1 5 - 6 1170 96
GenBank mRNA EU731009.1 - 4 - 5 621 96

View Full Product Details