Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD1D
Entrez Gene ID
100301530
Gene Name
CD1d molecule
Gene Aliases
-
Chromosome Location
Chr.13: 35069514 - 35074534 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160282.1 NP_001153754.1 1 - 2 206 80
XM_008264005.2 XP_008262227.1 2 - 3 648 80
GenBank mRNA FJ028656.1 - 1 - 2 206 80

View Full Product Details