Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD1D
Entrez Gene ID
100301530
Gene Name
CD1d molecule
Gene Aliases
-
Chromosome Location
Chr.13: 35069514 - 35074534 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160282.1 NP_001153754.1 3 - 4 761 104
XM_008264005.2 XP_008262227.1 4 - 5 1203 104
GenBank mRNA FJ028656.1 - 3 - 4 761 104

View Full Product Details