Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IFNG
Entrez Gene ID
100008602
Gene Name
interferon gamma
Gene Aliases
-
Chromosome Location
Chr.12: 101894547 - 101895994 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081991.1 NP_001075460.1 - 1053 90
GenBank mRNA AB010386.2 - - 1053 90
AF244933.1 - 2 - 3 182 90
DQ680162.1 - 2 - 3 182 90
DQ852341.1 - 2 - 3 182 90
EF589139.1 - 2 - 3 182 90
EF589140.1 - 2 - 3 182 90
EF589141.1 - 2 - 3 182 90

View Full Product Details