Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHOC
Entrez Gene ID
100008954
Gene Name
ras homolog family member C
Gene Aliases
-
Chromosome Location
Chr.13: 51451862 - 51457580 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002715765.4 XP_002715811.1 3 - 4 389 105
GenBank mRNA AY644387.1 - 2 - 3 245 105

View Full Product Details