Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GUCY2C
Entrez Gene ID
100008740
Gene Name
guanylate cyclase 2C
Gene Aliases
GCC
Chromosome Location
Chr.8: 24850182 - 24946807 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008259599.2 XP_008257821.1 17 - 18 3083 100
XM_017343367.1 XP_017198856.1 17 - 18 2447 100
GenBank mRNA AF184165.1 - 6 - 7 549 100

View Full Product Details