Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CACNB3
Entrez Gene ID
100009402
Gene Name
calcium voltage-gated channel auxiliary subunit beta 3
Gene Aliases
CAB3
Chromosome Location
Chr.4: 33141662 - 33146488 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101715.2 NP_001095185.1 11 - 12 1300 79
GenBank mRNA X64300.1 - - 1300 79

View Full Product Details