Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCGB1D
Entrez Gene ID
100008910
Gene Name
lipophilin BL
Gene Aliases
-
Chromosome Location
Chr.Unknown: 2366163 - 2369564 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082157.2 NP_001075626.1 - 122 64
GenBank mRNA AF308616.1 - - 57 64
AY303698.1 - - 123 64

View Full Product Details