Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COX6A1
Entrez Gene ID
100327270
Gene Name
cytochrome c oxidase subunit VIa
Gene Aliases
COX6A
Chromosome Location
Chr.21: 12132318 - 12134946 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170883.2 NP_001164354.1 1 - 2 143 70
GenBank mRNA AF198089.1 - 1 - 2 147 70

View Full Product Details