Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASIC4
Entrez Gene ID
100216510
Gene Name
acid sensing ion channel subunit family member 4
Gene Aliases
ACCN4
Chromosome Location
Chr.7: 160709687 - 160729807 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171017.1 NP_001164488.1 9 - 10 1505 77

View Full Product Details