Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTNNA1
Entrez Gene ID
100328952
Gene Name
catenin alpha 1
Gene Aliases
-
Chromosome Location
Chr.3: 21032448 - 21172627 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171142.1 NP_001164613.1 16 - 17 2435 82
XM_008254892.2 XP_008253114.1 16 - 17 2444 82

View Full Product Details